Liquid sorbent pads and pillows, Spilfyter® Kollect-A-Kem®
Supplier: NPS
Certificates
About this item
Use these sorbent materials in cleanrooms, laboratories, or anywhere chemical collection or absorption is needed. Material will not disintegrate and can be burned, leaving little ash. Two ply pads.
- Soaks up eight to ten times own weight in most tough chemicals
- Pillows eliminate the messy problems of expanded clay and other sandy, gritty material
- Low-linting polypropylene fabric rapidly collects most chemicals, acids, even water and hydrofluoric acid
Frequently Bought Together