Order Entry
Ireland
ContactUsLinkComponent
Chemical binder for spilled liquids, UNI-SAFE®
Chemical binder for spilled liquids, UNI-SAFE®
Catalog # 116-0000
undefined
Chemical binder for spilled liquids, UNI-SAFE®
Catalog # 116-0000
Specifications
  • Description:
    Chemical binder, UNI-SAFE®, tub
  • Pk:
    5 kg
Specifications
About this item
A universal oil and chemical binder safely absorbs all liquids without restriction and within a few minutes. Shows no dangerous reaction, not even with strongly reacting and oxidising substances such as nitric acid, oleum, hydrofluoric acid, bromine, sulphuric acid or hydrogen peroxide. Acts as an emission blocker by creating a steam barrier that protects both personnel and the environment. Once in contact with acid spillages, the binder changes from light green to yellow, where as with alkaline substances the green becomes darker.
  • Absorb up to 75% of its own weight (200 g absorbs 150 ml)
  • Safe enough to be applied by hand
  • Saturated substances can safely be swept, shovelled or pumped away