Order Entry
Ireland
ContactUsLinkComponent
Absorbent powder, Chemizorb® Hydrofluoric Acid (HF)
Absorbent powder, Chemizorb® Hydrofluoric Acid (HF)
Catalog # 1.01591.1000
Supplier:  Merck
undefined
Absorbent powder, Chemizorb® Hydrofluoric Acid (HF)
Catalog # 1.01591.1000
Supplier:  Merck
Specifications
  • Description:
    Absorbent powder, Chemizorb® HF
  • Packed:
    Bottle
  • Pk:
    1 kg
Specifications
About this item
Chemizorb Hydrofluoric Acid (HF) is a powder mixture consisting of a synthetic copolymer as the absorbent, a calcium salt as a neutraliser and precipitant, and a pH indicator.
  • Easy to use
  • pH indicator makes it possible to monitor the neutralisation process
  • 150 g of Chemizorb HF is required to absorb and neutralise 100 ml of hydrofluoric acid
The neutralisation process results in the release of CO₂. When the hydrofluoric acid has been neutralised, the colour of the indicator changes from red to orange/yellow.