Capillary electrophoresis tubing
Manufactured from precision bore tubing.
- Hydrophobic and hydrophilic surface treatments
- Wide pH range
- 30, 50, 75 and 100 µm Int.Ø
- Column-to-column reproducibility
Frequently Bought Together
Manufactured from precision bore tubing.