
4-Phenyl-1,2,3-thiadiazole ≥98%
Catalog # CAYM10011055-250
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:162.22 g/mol
EINECS:636-735-4
:GHS07
Storage Temperature:Freezer
CAS Number:25445-77-6
Formula:C₈H₆N₂S
MDL Number:MFCD00087232