Order Entry
Ireland
ContactUsLinkComponent
Ammonium standard solution, 500 mg/L NH₄ in water standard for Ion Chromatography
Ammonium standard solution, 500 mg/L NH₄ in water standard for Ion Chromatography
  YSIL7172
 :  
Ammonium standard solution, 500 mg/L NH₄ in water standard for Ion Chromatography
  YSIL7172
 :  

 

  • Concentration:
    500 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    150 mL
  • Description:
    Ammonium standard solution