Order Entry
Ireland
ContactUsLinkComponent
Formic acid 0,1%
Formic acid 0,1%
Catalog # ROTHCP03.1
Supplier:  Roth Carl
CAS Number:  
undefined
Formic acid 0,1%
Catalog # ROTHCP03.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :46.03 g/mol
Storage Temperature:Ambient
EINECS:200-579-1
REACH:01-2119491174-37-XXXX
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297