Tubings, PFA, BOLA
Supplier: Bohlender
Certificates
About this item
Made of PFA, transparent
- Temperature resistance: −200 to +260°C
- Gastighttubing
- Almost universal chemical resistance
- Resistant to irradiation and weather
- Sterilized in autoclaves
- Mechanical strength even at high temperatures
Applications include perfect tubing for aggressive and pure liquids or gases. Good bending radius and burst pressure.
Frequently Bought Together