Magnetic stirring bars, cylindrical, PTFE, BOLA
Supplier: Bohlender
Certificates
About this item
Standard magnetic stirring bar with PTFE-encapsulated magnetic core (Alnico 5).
- Universal chemical resistance
- Temperature resistance from –200 to +250 °C
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
Frequently Bought Together