Order Entry
Ireland
ContactUsLinkComponent
Test strips, zinc, QUANTOFIX®
Test strips, zinc, QUANTOFIX®
Catalog # 705-0311
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, zinc, QUANTOFIX®
Catalog # 705-0311
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Dithizon
  • Pk:
    100 Tests
  • Description:
    Zinc
  • Seq:
    38
  • Range:
    0 - 100 mg/l Zn₂+

Specifications

About this item

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips allow the quick and easy determination of Zinc in solutions. They come with all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 2 minutes.

Zinc provides an effective protective coating for steel (galvanised coatings) and is useful as an alloying agent. Zinc tests are used to regularly check the plating baths in metal plating industry. Zinc salts are useful as corrosion inhibitors in cooling water treatment formulations. Here, the zinc concentration is checked to ensure an optimal process.

Gradation: 0 · 2 · 5 · 10 · 25 · 50 · 100 mg/L Zn₂+

Colour: orange → red