Order Entry
Ireland
ContactUsLinkComponent
Visual colour kits, nickel, MQuant®, Supelco®
Visual colour kits, nickel, MQuant®, Supelco®
Catalog # 1.14420.0001
Supplier:  Merck
CAS Number:  
Visual colour kits, nickel, MQuant®, Supelco®
Catalog # 1.14420.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Chemical method:
    Dimethylglyoxime
  • Pk:
    125 Tests
  • Description:
    Nickel test
  • M-Clarity quality level:
    MQ100
  • Type:
    MCC
  • Range:
    0,02-0,04-0,07-0,1-0,15-0,2-0,3-0,4-0,5 mg/l Ni

Specifications

About this item

Quick tests, visual colour comparison.

  • Results in approximately 5 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available

MQuant® with colour card comparator:

By using cuvettes with a long light path, the MQuant® (MTC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/L). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.

Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.