Supelco® PESTANAL® Organic Reference Standard, Dalapon
35562-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:75-99-0
- Description:Dalapon
- Storage conditions:2...30 °C
- Hazard class description:H302 - H315 - H318 - H412
- Shipping conditions:STD
- MDL No.:MFCD00004178
- M-Clarity quality level:MQ100
- Synonym(s):2,2-Dichloropropionic acid; Dalapon
- Formula:CH₃CCl₂CO₂H
- Molecular weight:142,97
- Density:1,389 g/ml at 25 °C (lit.)
- Flash point:113 °C - closed cup
- Boiling point:90...92 °C/14 mm Hg (lit.)
- GHS signal: GHS05
: P264 - P273 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338