Palmitoleic acid
Supplier: Cayman Chemical
Jump to:
About this item
Synonyms: (Z)-Hexadec-9-enoic acid
- CAS Number:373-49-9
- Formula:C₁₆H₃₀O₂
- MDL Number:MFCD00004437
- Molecular Weight:254.41 g/mol
- EINECS:206-765-9
4 Options Available Below
Product Details & Documents
Synonyms: (Z)-Hexadec-9-enoic acid
- CAS Number:373-49-9
- Formula:C₁₆H₃₀O₂
- MDL Number:MFCD00004437
- Molecular Weight:254.41 g/mol
- EINECS:206-765-9
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice



