

tert-Amyl methyl ether analytical standard, Supelco®
Catalog # 442794.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:102.17 g/mol
Boiling Pt:85…86 °C (lit.)
Density:0,76…0.78 g/mL (25 °C) (lit.)
Flash Pt:–7 °C
EINECS:213-611-4
UN:3271
ADR:3,II
:GHS02
:GHS07
CAS Number:994-05-8
Formula:C₆H₁₄O
MDL Number:MFCD00010186