Organic reference standard, Dichlorodifluoromethane 200 µg/ml in methanol
Catalog # 123502W
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:75-71-8
- Description:Dichlorodifluoromethane 200 µg/ml in methanol
- Formula:CCl₂F₂
- Description:Dichlorodifluoromethane