Order Entry
Ireland
ContactUsLinkComponent
(±)-3-Chloro-1,2-propanediol, Supelco®
(±)-3-Chloro-1,2-propanediol, Supelco®
Catalog # 32406-100MG
Supplier:  Merck
CAS Number:  
undefined
(±)-3-Chloro-1,2-propanediol, Supelco®
Catalog # 32406-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :110.54 g/mol
Merck Index:13,02164
Boiling Pt:213 °C (1013 hPa)
Melting Pt:–40 °C
Density:1,32 g/cm³ (20 °C)
Flash Pt:135 °C
Storage Temperature:Ambient
EINECS:202-492-4
UN:2689
ADR:6.1,III
CAS Number:96-24-2
Formula:C₃H₇ClO₂
MDL Number:MFCD00004712