
2-Phenylpyridine
Catalog # MOLEM12155905
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:155.2 g/mol
Boiling Pt:269…270 °C (1013 hPa)
Melting Pt:76…77 °C
Density:1,623 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-763-1
UN:2810
ADR:6.1,III
:GHS07
CAS Number:1008-89-5
Formula:C₁₁H₉N
MDL Number:MFCD00006280