Order Entry
Ireland
ContactUsLinkComponent
2,4,6-Collidine for synthesis, Sigma-Aldrich®
2,4,6-Collidine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2,4,6-Collidine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    35 g/l (20 °C)
  • Melting Point:
    -43 °C
  • Molar Mass:
    121.18 g/mol
  • Boiling Point:
    170 - 172 °C (1013 hPa)
  • Vapor Pressure:
    4 hPa (20 °C)
  • Flash Point:
    55 °C
  • Refractive Index:
    1.50 (20 °C)
  • Density:
    0.914 g/cm3 (20 °C)
  • pH:
    (H2O) strongly alkaline

Specification Test Results

Related Information

Frequently Bought Together