Osmium tetraoxide
Catalog # MOLE11841630-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.23 g/mol
Merck Index:13,06962
Boiling Pt:130 °C (1013 hPa)
Melting Pt:40 °C
Density:1,04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:244-058-7
UN:2471
ADR:6.1,I
:GHS06
:GHS05
CAS Number:20816-12-0
Formula:O₄Os
MDL Number:MFCD00011150