
Zinc oxide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-13-2
:
Formula:ZnO
MDL Number:MFCD00011300
:81.39 g/mol
Merck Index:14,10147
Melting Pt:1975 °C
Density:5,606
Storage Temperature:Ambient
EINECS:215-222-5
UN:3077
ADR:9,III
REACH:01-2119463881-32
:GHS09
Frequently Bought Together