Order Entry
Ireland
ContactUsLinkComponent
1-Indanone
1-Indanone
Catalog # MOLEM33070742
Supplier:  Molekula
CAS Number:  
1-Indanone
Catalog # MOLEM33070742
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :132.16 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:37…40 °C
Density:1,1028 g/cm³ (40 °C)
Storage Temperature:Ambient
EINECS:201-470-1
CAS Number:83-33-0
Formula:C₉H₈O
MDL Number:MFCD00003785