
1-Indanone 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:83-33-0
:
Formula:C₉H₈O
MDL Number:MFCD00003785
:132.16 g/mol
Boiling Pt:243…245 °C (760 mmHg)
Melting Pt:38…42 °C
Density:1,1028 g/cm³ (40 °C)
Flash Pt:>110 °C
EINECS:201-470-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to brown Crystalline mass
- Infrared spectrum:Conforms
- Melting point:38 to 42 °C
- GC:≥99.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together