Order Entry
Ireland
ContactUsLinkComponent
Selenium dioxide 99.8% (trace metals basis)
Toxic
Health
Environment
Selenium dioxide 99.8% (trace metals basis)
Certificates
undefined
Toxic
Health
Environment
Selenium dioxide 99.8% (trace metals basis)

Specification Test Results

  • Appearance:
    White to light beige Powder
  • Assay:
    70 to 71.5 % Se
  • Total trace metal impurities:
    ≤0.2 %
  • Arsenic (As):
    ≤20 ppm
  • Cadmium (Cd):
    ≤5 ppm
  • Copper (Cu):
    ≤10 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm
  • Mercury (Hg):
    ≤5 ppm
  • Nickel (Ni):
    ≤10 ppm
  • Tellurium (Te):
    ≤500 ppm
  • Insoluble matter:
    ≤0.2 %

Specification Test Results

Related Information

Frequently Bought Together