

Hydrochloric acid 6.0 N, Biotech Reagent, J.T.Baker®
Catalog # 0347-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance (Clear, colorless liquid):Passes Test
- Normality:5.95 - 6.05
- Ammonium (NH₄):≤ 3 ppm
- Trace Impurities - Heavy Metals:≤ 0.5 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Sulfate (SO₄):≤ 1 ppm
Specification Test Results
About this item
:
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Density:1,06 g/cm³
ADR:8,II
:Management of Change (MOC) category = R
:36.46 g/mol
UN:1789
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324