
Germanium(IV) chloride ≥99.9999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10038-98-9
:
Formula:Cl₄Ge
MDL Number:MFCD00011029
:214.42 g/mol
Merck Index:14,4410
Boiling Pt:83,1 °C
Melting Pt:–49,5 °C
Density:1,874
Storage Temperature:Ambient
EINECS:233-116-7
UN:3264
ADR:8,II
:GHS05
Related Information
Frequently Bought Together