4-Hydrazinobenzoic acid
Catalog # MOLE13488749-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Hydrazinylbenzoic acid
:
:152.15 g/mol
EINECS:210-609-5
CAS Number:619-67-0
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007581