Order Entry
Ireland
ContactUsLinkComponent
Bis(cyclopentadienyl)titanium dichloride
Bis(cyclopentadienyl)titanium dichloride
Catalog # MOLEM83725447
Supplier:  Molekula
CAS Number:  
Bis(cyclopentadienyl)titanium dichloride
Catalog # MOLEM83725447
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride
 :
 :248.97 g/mol
EINECS:215-035-9
UN:3261
ADR:8,II
CAS Number:1271-19-8
Formula:C₁₀H₁₀Cl₂Ti
MDL Number:MFCD00003723