Order Entry
Ireland
ContactUsLinkComponent
 
 
Methacrylic acid alkylester (mixture of C₁₂-C₁₆) stabilised for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Methacrylic acid alkylester (mixture of C₁₂-C₁₆) stabilised for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -25 °C
  • Boiling Point:
    300 °C
  • Flash Point:
    105 °C
  • Density:
    0.87 g/cm3 (20 °C)
  • assay n-C12 isomer (GC, area%):
    60.0 - 70.0 % (a/a)

 

 

Frequently Bought Together