cis-1,2,3,6-Tetrahydrophthalic anhydride 95%
Catalog # B21431.30
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
Specifications
About this item
Synonyms: cis-4-Cyclohexene-1,2-dicarboxylic anhydride
CAS Number:935-79-5
:
Formula:C₈H₈O₃
:152.15 g/mol
Melting Pt:97…103 °C
EINECS:213-308-7
:GHS08
:GHS05
Storage Temperature:Ambient
MDL Number:MFCD00005916