N,N-Dimethylacetamide dimethyl acetal 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18871-66-4
:
Formula:C₆H₁₅NO₂
MDL Number:MFCD00008476
:133.19 g/mol
Boiling Pt:110…118 °C (760 mmHg)
Density:0,911 g/cm³
Flash Pt:8 °C
Storage Temperature:Ambient
EINECS:242-641-0
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4180 (20°C, 589 nm)
- Stabilizer:sodium carbonate
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together