Sodium sulphate
Supplier: Roth Carl
Certificates
About this item
CAS Number:7757-82-6
:
Formula:Na₂SO₄
MDL Number:MFCD00003504
Merck Index:12,08829
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:888 °C
Density:2,7 g/cm³ (20 °C)
:142.04 g/mol
Storage Temperature:Ambient
EINECS:231-820-9
Frequently Bought Together