Order Entry
Ireland
ContactUsLinkComponent
Nitric acid fuming ≥98% (by titrimetric analysis), Purum p.a.
 
 
 
Nitric acid fuming ≥98% (by titrimetric analysis), Purum p.a.
  84392-500ML
 :  
 
 
 
Nitric acid fuming ≥98% (by titrimetric analysis), Purum p.a.
  84392-500ML
 :  

 

  • Pack type:
    Glass bottle, coated
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Assay:
    Min. 98 %
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Total S (as SO₄):
    Max. 150 ppm
  • Chloride (Cl):
    Max. 10 ppm

 

 

 :
 :63.01 g/mol
 :13,06610
 :86 °C
 :–42 °C
 :1,52 g/cm³ (20 °C)
 :Ambient
 :231-714-2
 :2032
 :8,I
 :01-2119487297-23
 :GHS06
 :GHS05
 :GHS03
 :7697-37-2
 :HNO₃
 :MFCD00011349