Order Entry
Ireland
ContactUsLinkComponent
Sodium nitrite ≥98%, GPR RECTAPUR®
Oxidizing
Toxic
Environment
Sodium nitrite ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Toxic
Environment
Sodium nitrite ≥98%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 100 ppm
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together