
Sodium periodate
Catalog # HACH98499
Supplier: Hach
CAS Number:
Specifications
- Pk:100
- Seq:0100
Specifications
About this item
:
Merck Index:13,08714
Boiling Pt:300 °C (1013 hPa)
Melting Pt:175 °C
Density:3,87 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-197-6
UN:1479
ADR:5.1,II
:213.89 g/mol
:GHS03
CAS Number:7790-28-5
Formula:NaIO₄
MDL Number:MFCD00003534