Order Entry
Ireland
ContactUsLinkComponent
Potassium iodate 99.7-100.4%, Puriss. p.a., Fluka™
Oxidizing
Irritant
Potassium iodate 99.7-100.4%, Puriss. p.a., Fluka™
Certificates
Oxidizing
Irritant
Potassium iodate 99.7-100.4%, Puriss. p.a., Fluka™

Specification Test Results

  • Assay:
    99.7 - 100.4 %
  • Water insoluble matter:
    Max. 0.005 %
  • Loss on drying (130°C):
    Max. 0.05 %
  • pH (5 %, 25 °C):
    5.0 - 8.0
  • Copper (Cu):
    Max. 0.0005 %
  • Iron (Fe):
    Max. 0.001 %
  • Sodium (Na):
    Max. 0.005 %
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Chlorate, Chloride, Bromide, Bromate (as Cl):
    Max. 0.01 %
  • Total N:
    Max. 0.002 %
  • Iodide (I):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.005 %

Specification Test Results

Related Information

Frequently Bought Together