

Potassium iodate 98%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-05-6
:
Formula:KlO₃
MDL Number:MFCD00011406
:214 g/mol
Merck Index:13,07726
Melting Pt:560 °C
Density:3,93 g/cm³
Storage Temperature:Ambient
EINECS:231-831-9
UN:1479
ADR:5.1,II
:GHS07
:GHS03
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Titration Iodometric:≥97.5 %
- Loss on drying:≤0.5 % (105°C, 3 hrs)
Specification Test Results
Frequently Bought Together