

Sodium iodide, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7681-82-5
:
Formula:NaI
MDL Number:MFCD00003532
:149.89 g/mol
Merck Index:13,08705
Boiling Pt:1300 °C (1013 hPa)
Melting Pt:662 °C
Density:3,67 g/cm³ (20 °C)
UN:3077
ADR:9,III
:M-Clarity™ quality level = MQ300
EINECS:231-679-3
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1793 g/l (20 °C)
- Melting Point:662 °C
- Molar Mass:149.89 g/mol
- Bulk Density:1500 - 2000 kg/m3
- Boiling Point:1304 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (767 °C)
- Density:3.67 g/cm3 (20 °C)
- pH:6 - 9 (50 g/l, H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Frequently Bought Together