Potassium iodide, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Dipotassium diiodide, Kali iodide
CAS Number:7681-11-0
:
Formula:Kl
MDL Number:MFCD00011405
:166 g/mol
Merck Index:13,07727
Boiling Pt:1330 °C (1013 hPa)
Melting Pt:686 °C
Density:3,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-659-4
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:min. 99.0 %
- Chloride and Bromide (as Cl):max. 0.01 %
- Barium (Ba):max. 0.002 %
- Calcium (Ca):max. 0.002 %
- Magnesium (Mg):max. 0.001 %
- Sodium (Na):max. 0.005 %
- Insoluble Matter:max. 0.005 %
- Loss on Drying at 150°C:max. 0.2 %
- pH of 5% Solution at 25°C:6.0 - 9.2
- Phosphate (PO4):max. 0.001 %
- Sulfate (SO4):max. 0.005 %
- Iron (Fe):max. 3
- Heavy Metals (as Pb):max. 5
- Iodate (IO3):max. 3
Specification Test Results
Related Information
Frequently Bought Together