
Sodium fluoride ≥98.0% (fusing), Purum p.a.
Catalog # HONC71522-25KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay:98.0 - 102.0 %
- Calcium (Ca):max. 50 ppm
- Cadmium (Cd):max. 50 ppm
- Cobalt (Co):max. 50 ppm
- Copper (Cu):max. 50 ppm
- Iron (Fe):max. 50 ppm
- Potassium (K):max. 2000 ppm
- Nickel (Ni):max. 50 ppm
- Lead (Pb):max. 50 ppm
- Zinc (Zn):max. 50 ppm
- Total S (as SO₄):max. 200 ppm
Specification Test Results
About this item
:
:41.99 g/mol
Merck Index:13,08691
Boiling Pt:1,704 °C (1.013 hPa)
Melting Pt:990 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-667-8
UN:1690
ADR:6.1,III
:GHS06
CAS Number:7681-49-4
Formula:NaF
MDL Number:MFCD00003524