

Sodium fluoride ≥99% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-49-4
:
Formula:NaF
MDL Number:MFCD00003524
:41.99 g/mol
Merck Index:14,8618
Boiling Pt:1704 °C
Melting Pt:993 °C
Density:2,78
Storage Temperature:Ambient
EINECS:231-667-8
UN:1690
ADR:6.1,III
:GHS06
:GHS07
Related Information
Frequently Bought Together