Sodium chlorate 99+%, extra pure
:7775-09-9
:
:ClNaO₃
:MFCD00003479
:13,08670
:300 °C (1013 hPa)
:248…261 °C
:2,49 g/cm³
:Ambient
:231-887-4
:5.1,II
:106.44 g/mol
:1495
:01-2119474389-23
:GHS07
:GHS09
:GHS03
- Appearance:White to light yellow Crystalline powder or crystals
- Water:≤0.05 %
- Titration:≥99.0 %
- Sodium chloride (NaCl):≤1000 ppm
- Insoluble matter:≤0.005 % (in water)
Frequently Bought Together