VWR®, Filter Tips, Low Retention
Supplier: VWR Collection
Certificates
About this item
Low retention tips made of transparent PP resins offering unsurpassed fluid retention performance and proven aerosol blocking with endotoxin-free, 10 μm porous, hydrophobic filter. Ideal for PCR and DNA manipulation.
- Sustainable design based on virgin resins
- With PE filter, sterile
- Up to 10 times less fluid retention than standard tips
- Sample can be completely recovered without cutting into tip
- Lot-certified purity (free from protease, RNase/DNase, human DNA and endotoxin, non pyrogenic)
- Tips and racks are autoclavable to 121 °C for a maximum of 15 minutes (dry cycle is not recommended)
Specifications
- Catalog No:
- 732-1488
- 732-1490
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging