
Lithium carbonate
Catalog # MOLE13183442-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:73.89 g/mol
Merck Index:13,05549
Melting Pt:720 °C
Density:2,11 g/cm³ (20 °C)
EINECS:209-062-5
:GHS07
Storage Temperature:Ambient
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084