Order Entry
Ireland
ContactUsLinkComponent
Lithium carbonate ≥98.5%, pure, Supelco®
Irritant
Lithium carbonate ≥98.5%, pure, Supelco®
Catalog # 1.05670.9050
Supplier:  Merck
CAS Number:  
Irritant
Lithium carbonate ≥98.5%, pure, Supelco®
Catalog # 1.05670.9050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Fibre carton
  • Pk:
    50 kg
  • Seq:
    4418

Specifications

Specification Test Results

  • Assay (acidimetric):
    ≥ 98.5 %
  • Identity:
    passes test
  • Chloride (Cl):
    ≤ 0.05 %
  • Sulphate (SO₄):
    ≤ 0.4 %
  • Heavy metals (as Pb):
    ≤ 0.005 %
  • Ca (Calcium):
    ≤ 0.05 %
  • Fe (Iron):
    ≤ 0.003 %
  • K (Potassium):
    ≤ 0.02 %
  • Na (Sodium):
    ≤ 0.3 %

Specification Test Results

About this item

 :
 :73.89 g/mol
Merck Index:13,05549
Melting Pt:720 °C
Density:2,11 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:209-062-5
 :GHS07
Storage Temperature:Ambient
CAS Number:554-13-2
Formula:Li₂CO₃
MDL Number:MFCD00011084