Order Entry
Ireland
ContactUsLinkComponent
Potassium cyanide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Potassium cyanide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Potassium cyanide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    716 g/l (25 °C)
  • Melting Point:
    634 °C
  • Molar Mass:
    65.12 g/mol
  • Bulk Density:
    750 kg/m3
  • Boiling Point:
    1625 °C (1013 hPa)
  • Vapor Pressure:
    (20 °C)
  • Density:
    1.55 g/cm3 (20 °C)
  • pH:
    11 - 12 (20 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Related Information

Frequently Bought Together