Order Entry
Ireland
ContactUsLinkComponent
Potassium cyanide, Supelco®
Potassium cyanide, Supelco®
Catalog # 1.04965.1000
Supplier:  Merck
CAS Number:  
Potassium cyanide, Supelco®
Catalog # 1.04965.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :65.12 g/mol
Merck Index:13,07709
Boiling Pt:1625 °C (1013 hPa)
Melting Pt:634 °C
Density:1,55 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-792-3
UN:1680
ADR:6.1,I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :GHS09
CAS Number:151-50-8
Formula:KCN
MDL Number:MFCD00011397