Order Entry
Ireland
ContactUsLinkComponent
 
 
Potassium bromide ≥99.5% (by argentometric titration), Puriss. p.a. ACS, Fluka™
 
 
Potassium bromide ≥99.5% (by argentometric titration), Puriss. p.a. ACS, Fluka™
 

 

  • Assay:
    99.5 - 101.0 %
  • pH (5 %, 25 °C):
    5.0 - 8.8
  • Colour of the solution:
    complying
  • Clearness of the solution:
    complying
  • Loss on drying (130°C):
    Max. 0.3 %
  • Water insoluble matter:
    Max. 50 ppm
  • Arsenic (As):
    Max. 0.1 ppm
  • Barium (Ba):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Mercury (Hg):
    Max. 100 ppb
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 200 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Bromate (BrO₃):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 1000 ppm
  • Iodide (I):
    Max. 10 ppm
  • Iodate (IO₃):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 50 ppm
  • Total N:
    Max. 10 ppm

 

 

Frequently Bought Together