Order Entry
Ireland
ContactUsLinkComponent
Sodium metaarsenite ≥98%, GPR RECTAPUR®
Toxic
Environment
Sodium metaarsenite ≥98%, GPR RECTAPUR®
Catalog # 27687.265
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Environment
Sodium metaarsenite ≥98%, GPR RECTAPUR®
Catalog # 27687.265
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (Iodometric):
    Min. 98 %
  • Cl (Chloride):
    Max. 0.04 %
  • SO4 (Sulphate):
    Max. 0.4 %
  • Fe (Iron):
    Max. 0.022 %

Specification Test Results

About this item

 :
 :129.91 g/mol
Merck Index:13,08651
Melting Pt:615 °C
Density:1,87 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-070-5
UN:2027
ADR:6.1,II
 :GHS06
 :GHS09
CAS Number:7784-46-5
Formula:NaAsO₂
MDL Number:MFCD00003472