


1,4-Dioxane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:13,03330
Boiling Pt:101,1 °C (1013 hPa)
Melting Pt:11,8 °C
Density:1,03 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1165
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
EINECS:204-661-8
:GHS02
:GHS08
:GHS07
Specification Test Results
- Assay (on anhydrous substance):Min. 99.5 %
- Acidity or alkalinity:Max. 0.001 meq/g
- Boiling point:100 - 102 °C
- Colouration:Max. 10 APHA
- Density (20/4):1.030 - 1.035
- Solidification point:Min. 11 °C
- Acetaldehyde:Max. 40 ppm
- Acetone:Max. 5 ppm
- Evaporation residue:Max. 50 ppm
- Formaldehyde:Max. 5 ppm
- Peroxides:Max. 100 ppm
- Water:Max. 0.1 %
- Fe (Iron):Max. 1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC23540.295 spec_VWRC23540.295 spec_VWRC23540.295 spec_VWRC23540.320 spec_VWRC23540.320
Product specification
More
Frequently Bought Together