1-Ethynylcyclohexanol for synthesis, Sigma-Aldrich®
Catalog # 8.41522.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:10 g/l (20 °C)
- Melting Point:30 - 32 °C
- Molar Mass:124.18 g/mol
- Boiling Point:180 °C (1013 hPa)
- Vapor Pressure:1 hPa (50 °C)
- Flash Point:73 °C
- Explosion Limit:1.3 - 8.7 %(V)
- Density:0.976 g/cm3 (20 °C)
- pH:7 (1 g/l, H2O, 20 °C)
- Ignition Point:365 °C
Specification Test Results
About this item
Synonyms: 1-Ethynyl-1-cyclohexanol
:
:124.18 g/mol
Boiling Pt:180 °C (1013 hPa)
Melting Pt:30…32 °C
Density:0,973 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-100-9
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS06
CAS Number:78-27-3
Formula:C₈H₁₂O
MDL Number:MFCD00003858